Skip to Content
Merck
All Photos(1)

Key Documents

531677

Sigma-Aldrich

m-Tolyl acetate

97%

Synonym(s):

3-Methylphenylacetate, m-Cresyl acetate, Acetic acid m-tolyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OCOCH3
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.501 (lit.)

bp

210-213 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CC(=O)Oc1cccc(C)c1

InChI

1S/C9H10O2/c1-7-4-3-5-9(6-7)11-8(2)10/h3-6H,1-2H3

InChI key

OTGAHJPFNKQGAE-UHFFFAOYSA-N

General description

m-Tolyl acetate is an acyloxy benzene derivative that can be prepared from m-cresol and acetic anhydride.

Application

m-Tolyl acetate may be used in the preparation of 2-hydroxy-4-methylacetophenone and 4-hydroxy-2-methyl acetophenone via Fries rearrangement.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracanal medication in endodontic treatment: a survey of endodontic programs.
J M Gergely et al.
General dentistry, 41(4), 328-331 (1993-07-01)
[Basic studies on the clinical application of cresatin].
K Miyazawa
Shigaku = Odontology; journal of Nihon Dental College, 70(6), 1268-1279 (1983-04-01)
Synthesis and characterization of o-hydroxyarylalkylketones by using eco-friendly solvent free catalyst in frie's rearrangement.
Chouke PB and Vishwas NI.
Der Pharma Chemica, 4(1), 377-382 (2012)
Systemic effects of endodontic intracanal medicaments, irrigants and sealers: danger on the bracket table.
D R Morse et al.
Annals of dentistry, 44(1), 6-15 (1985-01-01)
P M DiFiore et al.
Oral surgery, oral medicine, and oral pathology, 55(1), 91-94 (1983-01-01)
Four calcium hydroxide-based apexification pastes were tested for their antibacterial effects on Streptococcus sanguis. Their zones of growth inhibition on blood agar plates were measured at 2, 4, 6, and 8 days. Only the camphorated parachlorophenol and the metacresylacetate pastes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service