Skip to Content
Merck
All Photos(1)

Key Documents

291390

Sigma-Aldrich

p-Tolyl acetate

99%

Synonym(s):

p-Cresyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4CH3
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.501 (lit.)

bp

210-211 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CC(=O)Oc1ccc(C)cc1

InChI

1S/C9H10O2/c1-7-3-5-9(6-4-7)11-8(2)10/h3-6H,1-2H3

InChI key

CDJJKTLOZJAGIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The Fries rearrangement of p-tolyl acetate has been investigated using the H-form of various zeolites as catalysts. Mechanism of photo-Fries rearrangement of p-tolyl acetate has been studied.

Application

p-Tolyl acetate has been used in the total synthesis of (−)-incrustoporin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q Yu et al.
Journal of Asian natural products research, 1(3), 183-188 (2001-03-20)
(-)-Incrustoporin (1) has been synthesized using aldol condensation of ethyl p-tolyl-acetate (2) and (2R)-benzoyloxy-butanal (3), followed by acid-catalyzed deprotection of the benzoyl group, lactone ring-closure, and elimination of the beta-OH in a one-pot manner. The aldehyde 3 was prepared from
Mechanistic studies of the photo-Fries reaction.
Sandner MR, et al.
Journal of the American Chemical Society, 90(26), 7249-7254 (1968)
Fries rearrangement over zeolitic catalysts.
Vogt A, et al.
Applied Catalysis A: General, 123(1), 37-49 (1995)
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Articles

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service