Skip to Content
Merck
All Photos(1)

Key Documents

528013

Sigma-Aldrich

Dimethyl disulfide

≥99%

Synonym(s):

DMDS, Methyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SSCH3
CAS Number:
Molecular Weight:
94.20
Beilstein:
1730824
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.24 (vs air)

Quality Level

vapor pressure

22 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

>572 °F

expl. lim.

16 %

availability

available only in EU

refractive index

n20/D 1.525 (lit.)

bp

109 °C (lit.)

mp

−85 °C (lit.)

density

1.046 g/mL at 25 °C (lit.)

functional group

disulfide

SMILES string

CSSC

InChI

1S/C2H6S2/c1-3-4-2/h1-2H3

InChI key

WQOXQRCZOLPYPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl disulfide can be used:
  • For the sulfidation of WS2 nanoparticles during the preparation of WS2/SBA15 mesoporous silica composite.
  • In the copper-catalyzed sulfenylation of 2,3-allenoic acids to afford 4-sulfenylated butenolides.
  • To synthesize various aryl sulfides by nickel-catalyzed thiolation of aryl iodides.
  • Along with triflic anhydride, as a promoter for glycosylation reaction.
  • In the copper-catalyzed 1,2-hydroxysulfenylation of alkenes to synthesize 1,2-acetoxysulfides.
  • In the synthesis of sulfur containing polycyclic aromatic hydrocarbons such as dibenzoannelated tetrathienoacene.

Legal Information

Product of Arkema

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Upper respiratory tract

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High loading of short WS2 slabs inside SBA-15: promotion with nickel and performance in hydrodesulfurization and hydrogenation.
Vradman L, et al.
J. Catal., 213(2), 163-175 (2003)
Copper-catalyzed 1, 2-hydroxysulfenylation of alkene using disulfide via cleavage of the s? s bond.
Taniguchi N
The Journal of Organic Chemistry, 71(20), 7874-7876 (2006)
A New, Powerful Glycosylation Method: Activation of Thioglycosides with Dimethyl Disulfide? Triflic Anhydride.
Tatai J and Fugedi P
Organic Letters, 9(22), 4647-4650 (2007)
Alkyl-or arylthiolation of aryl iodide via cleavage of the S? S bond of disulfide compound by Nickel Catalyst and Zinc.
Taniguchi N
The Journal of Organic Chemistry, 69(20), 6904-6906 (2004)
Dibenzoannelated tetrathienoacene: synthesis, characterization, and applications in organic field-effect transistors.
Huang J, et al.
Organic Letters, 14(13), 3300-3303 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service