Skip to Content
Merck
All Photos(3)

Key Documents

274380

Sigma-Aldrich

Dimethyl sulfide

anhydrous, ≥99.0%

Synonym(s):

DMS, Methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S
CAS Number:
Molecular Weight:
62.13
Beilstein:
1696847
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor density

2.1 (vs air)

vapor pressure

26.24 psi ( 55 °C)
7.79 psi ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

402 °F

expl. lim.

19.7 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.435 (lit.)

bp

38 °C (lit.)

mp

−98 °C (lit.)

solubility

H2O: soluble 7.28 g/L at 20 °C

density

0.846 g/mL at 25 °C (lit.)

SMILES string

CSC

InChI

1S/C2H6S/c1-3-2/h1-2H3

InChI key

QMMFVYPAHWMCMS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfide is the major biogenic, volatile sulfur compound that is released from the ocean to the atmosphere. It plays a main role in the global sulfur cycle.

Application

Dimethyl sulfide undergoes thiolation reaction with hydrogen sulfide (H2S) in the presence of tungsten-zirconia (WO3/ZrO2) catalyst to form methanethiol.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-32.8 °F - closed cup

Flash Point(C)

-36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photooxidation of dimethyl sulfide and dimethyl disulfide. I: Mechanism development
Yin F, et al
Journal of Atmospheric Chemistry, 11(4), 309-364 (1990)
Dimethyl sulfide production during natural phytoplanktonic blooms
Nguyen BC, et al.
Marine Chemistry, 24(2), 133-141 (1988)
Thiolation of dimethyl sulfide to methanethiol over WO3/ZrO2 catalysts
Chen S, et al
J. Mol. Catal. A: Chem., 365, 60-65 (2012)
Dimethyl sulfide production during natural phytoplanktonic blooms.
Nguyen BC, et al.
Marine Chemistry, 24(2), 133-141 (1988)
Ingrid Elisia et al.
PloS one, 11(3), e0152538-e0152538 (2016-04-01)
Dimethyl sulfoxide (DMSO) is currently used as an alternative treatment for various inflammatory conditions as well as for cancer. Despite its widespread use, there is a paucity of data regarding its safety and efficacy as well as its mechanism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service