E26223
Diethyl disulfide
99%
Synonym(s):
Ethyl disulfide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(C2H5)2S2
CAS Number:
Molecular Weight:
122.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
refractive index
n20/D 1.506 (lit.)
bp
151-153 °C (lit.)
density
0.993 g/mL at 25 °C (lit.)
SMILES string
CCSSCC
InChI
1S/C4H10S2/c1-3-5-6-4-2/h3-4H2,1-2H3
InChI key
CETBSQOFQKLHHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Diethyl Disulfide is a chemical reactant andindustrial catalyst with significant catalytic properties. It is a compoundthat participates in solid-gas reactions, particularly with iron foils at higher temperatures, leading to the formation of iron sulfide films.It can react with hydrogen at 200°C to produce ethanethiol, exhibiting higher reactivity and selectivity compared to reactions in helium.
Application
- Pheniramine maleate in forensic toxicology: Explored the assessment of pheniramine maleate in alternative biological matrices using liquid chromatography tandem mass spectrometry (LC-MS/MS). This research underscores the importance of pheniramine maleate in toxicological studies, contributing to forensic investigations and therapeutic drug monitoring (Makhdoom et al., 2024).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
104.0 °F - closed cup
Flash Point(C)
40 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of iron sulfide films through solid-gas reaction of iron with diethyl disulfide
Ocampo-Macias, T and Lara-Romero, et al.
Journal of Sulfur Chemistry, 36, 385-394 (2015)
Selective hydrogenation of diethyl disulfide to ethanethiol in the presence of sulfide catalysts
Mashkina, AV and Khairulina, et al.
Kinetics and Catalysis, 43, 261-267 (2002)
Bofan Yu et al.
Journal of chromatography. A, 1356, 221-229 (2014-07-07)
For the expanded application area, fast trace analysis of certain high boiling point (i.e., 150-250 °C) volatile organic compounds (HVOCs) in water, a large volume-direct aqueous injection-gas chromatography (LV-DAI-GC) method was optimized for the following parameters: packed sorbent for sample
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service