Skip to Content
Merck
All Photos(3)

Key Documents

647098

Sigma-Aldrich

Phenylboronic acid pinacol ester

97%

Synonym(s):

(Pinacolboryl)benzene, 4,4,5,5-Tetramethyl-2-phenyl-1,3,2-dioxaborolane, Pinacol phenylboronate, cyclic tetramethylethylene ester benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17BO2
CAS Number:
Molecular Weight:
204.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

27-31 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2ccccc2

InChI

1S/C12H17BO2/c1-11(2)12(3,4)15-13(14-11)10-8-6-5-7-9-10/h5-9H,1-4H3

InChI key

KKLCYBZPQDOFQK-UHFFFAOYSA-N

General description

Phenylboronic acid, pinacol ester, also known as boronate ester, is generally used in metal-catalyzed C-C bond formation reactions like Suzuki–Miyaura reaction.

Application

Phenylboronic acid pinacol ester can be used:
  • To prepare sulfinamide derivatives by reacting with diethylaminosulfur trifluoride (DAST) and potassium phenyltrifluoroborate.
  • As a substrate in the study of Suzuki–Miyaura coupling of various aryl iodides over SiliaCat Pd(0).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-Free Cross-Coupling of Arylboronic Acids and Derivatives with DAST-Type Reagents for Direct Access to Diverse Aromatic Sulfinamides and Sulfonamides
Wang Q, et al.
Angewandte Chemie (International Edition in English), 55(36), 10811-10815 (2016)
Heterogeneous Pd/C-catalyzed ligand-free Suzuki-Miyaura coupling reaction using aryl boronic esters
Kitamura Y, et al.
Tetrahedron, 63(43), 10596-10602 (2007)
Enhanced heterogeneously catalyzed Suzuki-Miyaura reaction over SiliaCat Pd (0)
Pandarus V, et al.
Tetrahedron Letters, 54(35), 4712-4716 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service