Skip to Content
Merck
All Photos(1)

Key Documents

517747

Sigma-Aldrich

Aluminum oxide, mesoporous

MSU-X (wormhole), average pore size 3.8 nm

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

Quality Level

description

molecular sieve

form

powder

particle size

5.65 μm (avg.)

pore size

3.8 nm average pore size

application(s)

battery manufacturing

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1.018161.041611.01216
agency

ChP, NF

agency

NF

agency

ChP, NF

agency

-

form

solid

form

solid

form

solid

form

solid

bulk density

850 kg/m3

bulk density

-

bulk density

-

bulk density

850 kg/m3

solubility

754-764 g/L

solubility

754-764 g/L

solubility

-

solubility

754-764 g/L

product line

EMPROVE® EXPERT

product line

EMPROVE® EXPERT

product line

EMPROVE® EXPERT

product line

-

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    G Siddiqi et al.
    Dalton transactions (Cambridge, England : 2003), 44(32), 14349-14353 (2015-07-23)
    A simple one-step and gram-scale synthesis of [Au5Mes5] from AuCl3 was developed, and this molecular precursor was used to generate Au nanoparticles on SiO2 and Al2O3. While [Au5Mes5] does not react with surface silanols and is only physisorbed, its incipient
    Colin J Ingham et al.
    Biotechnology advances, 30(5), 1089-1099 (2011-08-23)
    Porous aluminum oxide (PAO) is a ceramic formed by an anodization process of pure aluminum that enables the controllable assembly of exceptionally dense and regular nanopores in a planar membrane. As a consequence, PAO has a high porosity, nanopores with
    Chien-Chih Lin et al.
    Nanoscale, 5(17), 8090-8097 (2013-07-25)
    We demonstrated a promising route for enhancing temperature sensitivity, improving saturation voltage, and reducing power consumption of the MOS(p) tunneling temperature sensors by introducing ultrathin Al2O3 into the dielectric stacks. Detailed illustrations of the working mechanism and device concept are
    Yi Tian et al.
    Journal of the American Chemical Society, 136(36), 12784-12793 (2014-08-19)
    Organic neutral radicals have long fascinated chemists with a fundamental understanding of structure-reactivity relationships in organic reactions and with applications as new functional materials. However, the elusive nature of these radicals makes the synthesis, isolation, and characterization very challenging. In
    Joohoon Kang et al.
    ACS nano, 9(4), 3596-3604 (2015-03-19)
    Solution dispersions of two-dimensional (2D) black phosphorus (BP)--often referred to as phosphorene--are achieved by solvent exfoliation. These pristine, electronic-grade BP dispersions are produced with anhydrous organic solvents in a sealed-tip ultrasonication system, which circumvents BP degradation that would otherwise occur

    Articles

    Mesoporous Materials include a range of high surface area porous silicates with applications in gas adsorption, drug delivery, diagnostics and catalysis.

    Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

    Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

    Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

    See All

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service