Skip to Content
Merck
All Photos(2)

Key Documents

486027

Sigma-Aldrich

L-Valine-d8

≥98 atom % D, ≥98% (CP)

Synonym(s):

(S)-α-Aminoisovaleric acid-2,3,4,4,4,5,5,5-d8, Deuterated L-valine, L-2-Amino-3-methylbutanoic acid-2,3,4,4,4,5,5,5-d8

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2CDCD(NH2)CO2H
CAS Number:
Molecular Weight:
125.20
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

solid

optical activity

[α]20/D +27.5°, c = 8 in 6 M HCl

mp

295-300 °C (subl.) (lit.)

mass shift

M+8

storage temp.

room temp

SMILES string

[2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])[C@]([2H])(N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i1D3,2D3,3D,4D

InChI key

KZSNJWFQEVHDMF-AYWPRJOCSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan M Marron et al.
Metabolites, 9(5) (2019-05-06)
Black versus white older Americans are more likely to experience frailty, a condition associated with adverse health outcomes. To reduce racial disparities in health, a complete understanding of the pathophysiology of frailty is needed. Metabolomics may further our understanding by
Haoxin Li et al.
Nature medicine, 25(5), 850-860 (2019-05-10)
Despite considerable efforts to identify cancer metabolic alterations that might unveil druggable vulnerabilities, systematic characterizations of metabolism as it relates to functional genomic features and associated dependencies remain uncommon. To further understand the metabolic diversity of cancer, we profiled 225
Andrew J Chetwynd et al.
Small (Weinheim an der Bergstrasse, Germany), 16(21), e2000295-e2000295 (2020-04-03)
Nanomaterials (NMs) are promptly coated with biomolecules in biological systems leading to the formation of the so-called corona. To date, research has predominantly focused on the protein corona and how it affects NM uptake, distribution, and bioactivity by conferring a
Eric A Franzosa et al.
Nature microbiology, 4(2), 293-305 (2018-12-12)
The inflammatory bowel diseases (IBDs), which include Crohn's disease (CD) and ulcerative colitis (UC), are multifactorial chronic conditions of the gastrointestinal tract. While IBD has been associated with dramatic changes in the gut microbiota, changes in the gut metabolome-the molecular
Mohamad-Reza Aghanoori et al.
Molecular metabolism, 20, 149-165 (2018-12-14)
Diabetic sensorimotor polyneuropathy (DSPN) affects approximately half of diabetic patients leading to significant morbidity. There is impaired neurotrophic growth factor signaling, AMP-activated protein kinase (AMPK) activity and mitochondrial function in dorsal root ganglia (DRG) of animal models of type 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service