488917
DL-Alanine-2,3,3,3-d4
98 atom % D
Synonym(s):
(±)-2-Aminopropionic acid-2,3,3,3-D4, Deuterated DL-alanine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CD3CD(NH2)CO2H
CAS Number:
Molecular Weight:
93.12
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
98 atom % D
Assay
99% (CP)
form
solid
mp
289 °C (dec.) (lit.)
mass shift
M+4
SMILES string
[2H]C([2H])([2H])C([2H])(N)C(O)=O
InChI
1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/i1D3,2D
InChI key
QNAYBMKLOCPYGJ-MZCSYVLQSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Renyu Zhang et al.
Food chemistry, 347, 128999-128999 (2021-01-20)
The effect of in-bag dry- and wet-ageing on metabolite profiles of lamb legs was determined using Rapid Evaporative Ionisation Mass Spectrometry (REIMS). Using orthogonal projection to latent structures-discriminant analysis (OPLS-DA) with REIMS, 1705 metabolite ions were identified (Q2 = 0.86) in four
Ren Kimura et al.
Scientific reports, 10(1), 804-804 (2020-01-23)
The incidence of dementia, a clinical symptom characterized by severe cognitive decline, is increasing worldwide. Predictive biomarkers are therefore required for early identification and management. D-amino acids in the brain contribute to cognitive function and are suggested as useful biomarkers
Yutaka Umakoshi et al.
Journal of bioscience and bioengineering, 128(6), 773-779 (2019-07-14)
Although d-amino acids are less prevalent in nature, they have been detected in mammals (including humans) and it is widely accepted that they might play important physiological roles. While an analytical method for chiral amino acid profiling is strongly required
M S Brook et al.
American journal of physiology. Endocrinology and metabolism, 313(6), E681-E689 (2017-08-16)
Current methods to quantify in vivo RNA dynamics are limited. Here, we developed a novel stable isotope (D
Yutaka Konya et al.
Journal of bioscience and bioengineering, 123(1), 126-133 (2016-09-21)
Recently, the demand for d-amino acid profiling has been drastically increasing because the significance of d-amino acid in various biological events is suggested. However, the present methodologies for d-amino acid profiling are still unsatisfactory. Therefore, a highly sensitive, robust, high-throughput
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service