Skip to Content
Merck
All Photos(1)

Key Documents

616192

Sigma-Aldrich

L-Lysine-4,4,5,5-d4 hydrochloride

98 atom % D, 98% (CP)

Synonym(s):

SILAC Amino Acid, (S)-2,6-Diaminocaproic acid-4,4,5,5-d4 hydrochloride, Deuterated Lys, Deuterated lysine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2(CD2)2CH2CH(NH2)CO2H · HCl
CAS Number:
Molecular Weight:
186.67
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

solid

optical activity

[α]25/D +20.7°, c = 2 in 5 M HCl

technique(s)

protein expression: suitable

mp

263-264 °C (dec.) (lit.)

mass shift

M+4

SMILES string

Cl.[2H]C([2H])(CN)C([2H])([2H])C[C@H](N)C(O)=O

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1/i1D2,2D2;

InChI key

BVHLGVCQOALMSV-UGJIAQRQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Sedigh Jasour et al.
Journal of agricultural and food chemistry, 65(48), 10673-10684 (2017-11-10)
The effect of partially replacing fishmeal in aquafeed with feathermeal (FTH) at three levels (0%: FTH0, 8%: FTH8, 24%: FTH24) and two extrusion temperatures (100 and 130 °C) was evaluated in rainbow trout (Oncorhynchus mykiss) with respect to growth performance
Zhifei Chen et al.
Journal of agricultural and food chemistry, 67(19), 5634-5646 (2019-04-25)
Proteins present in infant formulas are modified by oxidation and glycation during processing. Modified amino acid residues released from proteins may be absorbed in the gastrointestinal tract, and pose a health risk to infants. In this study, the markers of
Flavia Bianchi et al.
Insects, 11(11) (2020-11-22)
The production of phagostimulant and attractive volatile organic compounds (VOCs) by yeasts can be exploited to improve the efficacy of attract-and-kill formulations against the spotted wing drosophila (SWD). This study evaluated the persistence over one week of a yeast-based formulation
Zehan Hu et al.
Cell reports, 28(13), 3486-3496 (2019-09-26)
The target of rapamycin complex 1 (TORC1) is a master regulator of cell homeostasis, which promotes anabolic reactions and synchronously inhibits catabolic processes such as autophagy-mediated protein degradation. Its prime autophagy target is Atg13, a subunit of the Atg1 kinase
Zhifei Chen et al.
Free radical research, 53(1), 68-81 (2019-01-17)
Modification of proteins in infant milk formula (IF) is of major concern to the dairy industry and consumers. Thermal treatment is required for microbiological safety, but heat, light, metal-ions and other factors may induce oxidative damage, and be a health

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service