Skip to Content
Merck
All Photos(3)

Key Documents

483893

Sigma-Aldrich

N,N-Dimethylmethyleneiminium chloride

technical grade, 90%

Synonym(s):

Dimethylformiminium chloride, Methylenedimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=N+(CH3)2Cl-
CAS Number:
Molecular Weight:
93.56
Beilstein:
505955
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

mp

146-148 °C (lit.)

SMILES string

[Cl-].C[N+](C)=C

InChI

1S/C3H8N.ClH/c1-4(2)3;/h1H2,2-3H3;1H/q+1;/p-1

InChI key

ZJTROANVDZIEGB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reacant for:
Preparation of dihydronaphthyridinones as HIV-1 integrase inhibitors
Mannich reactions
Preparation of cyanotetrahydrooxo-ß-carbolines via cyclocondensation reactions
Asymmetric synthesis of phalarine via stereospecific Pictet-Spengler cyclocondensation and traceless chirality transfer from L-tryptophan
Synthesis of functionalized diarylpyrrolizines as inhibitors of microsomal prostaglandin E2 synthase-1 (mPGES-1) and 5-lipoxygenase (5-LOX)
Synthesis of (±)-phalarine with the rearrangement of an azaspiroindolenine and Gassman oxindole preparation as key steps

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service