Skip to Content
Merck
All Photos(1)

Key Documents

47675

Sigma-Aldrich

Formaldehyde diethyl acetal

absolute, over molecular sieve (H2O ≤0.01%), ≥99.0% (GC)

Synonym(s):

Diethoxymethane, Ethylal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(OC2H5)2
CAS Number:
Molecular Weight:
104.15
Beilstein:
1697253
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥99.0% (GC)
bp:
87-88 °C (lit.)
vapor pressure:
60 mmHg ( 25 °C)

vapor density

3.6 (vs air)

Quality Level

vapor pressure

60 mmHg ( 25 °C)

grade

absolute

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

346 °F

quality

over molecular sieve (H2O ≤0.01%)

refractive index

n20/D 1.373 (lit.)
n20/D 1.374

bp

87-88 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCOCOCC

InChI

1S/C5H12O2/c1-3-6-5-7-4-2/h3-5H2,1-2H3

InChI key

KLKFAASOGCDTDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Formaldehyde diethyl acetal (FDEA, diethoxymethane, DEM) is a diethyl acetal. It has been synthesized from ethanol and aqueous formaldehyde. It is a low boiling solvent useful for sodium hydride reactions, organolithium chemistry, copper-catalyzed conjugate additions and phase-transfer reactions. It is a potential alternate for tetrahydrofuran, dichloromethane, glyme and methylal. DEM is an ethoxymethylating agent, a formaldehyde equivalent and a carbonylation substrate. Its condensation with 2-propylresorcinol to form resorcin[n]arenes (n=4–7) has been investigated. The IR and Raman spectra and conformations of DEM have been studied. NMR spectroscopy has been used to study the aggregation behavior of organolithium compounds in diethoxymethane. Unimolecular metastable decomposition of DEM upon electron impact has been studied. Solubility of non-polar gases in DEM has been evaluated.

Application

Formaldehyde diethyl acetal may be used as a solvent in the esterification of carboxylic acids and in the O-alkylation of a variety of phenols.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NMR spectroscopy of organolithium compounds, Part XVI: The aggregation behavior of butyllithium, phenyllithium, and lithium diisopropylamide in dimethoxy-and diethoxymethane.
Bergander K, et al.
Tetrahedron, 50(20), 5861-5868 (1994)
Sc(OTf)3-catalyzed cyclocondensation of 2-propylresorcinol with diethoxymethane. Formation and fragmentation of resorcin [n] arenes.
Morikawa O, et al.
Tetrahedron Letters, 45(29), 5731-5734 (2004)
Use of Diethoxymethane as a Solvent for Phase-Transfer Esterification of Carboxylic Acids.
Coleman MT.
Synthetic Communications, 42(13), 1911-1913 (2012)
Solubility of non polar gases in formaldehyde diethyl acetal between-10 and 30?C, and 1 Atm partial pressure of gas.
Lizano LP, et al.
Journal of Solution Chemistry, 19(7), 721-728 (1990)
Metastable decompositions of gem-dialkoxyalkanes upon electron impact. III. Diethoxymethane (CH2 (OCH2CH3)2).
Tajima S, et al.
Rapid Communications in Mass Spectrometry, 14(14), 1195-1199 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service