Skip to Content
Merck
All Photos(1)

Key Documents

689386

Sigma-Aldrich

Formaldehyde dibutyl acetal

puriss., ≥97.0% (GC)

Synonym(s):

Dibutyl formal, Butylal, Dibutoxymethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(OCH2CH2CH2CH3)2
CAS Number:
Molecular Weight:
160.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

puriss.

Quality Level

Assay

≥97.0% (GC)

form

liquid

impurities

≤0.25% water

refractive index

n20/D 1.406

density

0.835 g/mL at 20 °C

functional group

ether

SMILES string

CCCCOCOCCCC

InChI

1S/C9H20O2/c1-3-5-7-10-9-11-8-6-4-2/h3-9H2,1-2H3

InChI key

QLCJOAMJPCOIDI-UHFFFAOYSA-N

Related Categories

General description

Formaldehyde dibutyl acetal is an acetal used in the manufacture of synthetic resins, antiseptics, deodorants, and fungicides. It is also used as a fuel additive to increase the octane number of gasoline or the n-cetane number of diesel fuels and reduce smoke and particulate emissions.

Application

Formaldehyde dibutyl acetal is a halogen-free and less toxic solvent that can be used to solubilize commercial low-density polyethylene (LDPE) samples to analyze molecular weight distribution using gel permeation chromatography (GPC). It can also be used as a reactant to prepare butoxymethyltriphenylphosphonium iodide, which is used for carbon homologation and also as a useful key intermediate in organic synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selected physicochemical properties of dibutoxymethane
P-T Marcela and Szafranski AM
Journal of Chemical and Engineering Data, 45(6), 988-990 (2000)
Facile synthesis of α-alkoxymethyltriphenylphosphonium iodides: new application of PPh 3/I 2
Gondal HY, et al.
Chemistry Central Journal, 12(1), 1-10 (2018)
Characterization of low-density polyethylene in dibutoxymethane by high-temperature gel permeation chromatography with triple detection
Boborodea A, et al.
Chromatographia, 79(15), 971-976 (2016)
Vieille-Petit, L.; et al.
Chemical Communications (Cambridge, England), 3783-3783 (2009)
Xinjun Luan et al.
Journal of the American Chemical Society, 130(21), 6848-6858 (2008-05-01)
A new class of easily accessible and stable imidazolin-2-ylidenes has been synthesized where the side chains are comprised of substituted naphthyl units. Introduction of the naphthyl groups generates C 2 -symmetric ( rac) and C s- symmetric ( meso) atropisomers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service