Skip to Content
Merck
All Photos(1)

Key Documents

15512

Sigma-Aldrich

Formaldehyde solution

meets analytical specification of USP, ≥34.5 wt. %

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12352114
eCl@ss:
39021101
PubChem Substance ID:
NACRES:
NA.21

vapor density

1.03 (vs air)

Quality Level

vapor pressure

52 mmHg ( 37 °C)

autoignition temp.

572 °F

quality

meets analytical specification of USP

contains

9.0-15.0% methanol as stabilizer (GC)

concentration

≥34.5 wt. %
25-50%

impurities

acidic reac. substances, complies
residual solvents, complies

ign. residue

≤0.01% (as SO4)

density

1.09 g/mL at 25 °C (lit.)

suitability

corresponds for identity

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formaldehyde solution commercially formaldehyde solution is an aqueous solution containing 37% formaldehyde and 8-10% methanol. Formaldehyde solution can be activated by adding the catalytic amount of lanthanide triflate. This activated formaldehyde solution was employed for the smooth hydroxymethylation reaction of silyl enol ethers. Formaldehyde solution reacts with silyl enol ethers to afford the corresponding hydroxymethylated adducts in high yields.

Application

Formaldehyde solution was used to prepare the formaldehyde stock solution for the development of transparent sol-gels with entrapped sensitive and selective reagents for the detection of formaldehyde in air. It may be used in the preparation of nanocrystalline metal (Cu, Hg, Zn, Bi, Pb) sulfides having different shapes and different particle sizes.

Other Notes

The article number 15512-4X2.5L-R will be discontinued. Please order the single bottle 15512-2.5L-R which is physically identical with the same exact specifications.
The article number 15512-6X1L-R will be discontinued. Please order the single bottle 15512-1L-R which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lanthanide Trifluoromethanesulfonates as Stable Lewis Acids in Aqueous Media. Yb (OTf)3 Catalyzed Hydroxymethylation Reaction of Silyl Enol Ethers with Commercial Formaldehyde Solution.
Kobayashi S.
Chemistry Letters (Jpn), 12, 2187-2190 (1991)
Microwave synthesis of nanocrystalline metal sulfides in formaldehyde solution.
Liao X-H, et al.
Materials Science and Engineering, B, 85(1), 85-89 (2001)
Lanthanide triflates as water-tolerant Lewis acids. Activation of commercial formaldehyde solution and use in the aldol reaction of silyl enol ethers with aldehydes in aqueous media.
Kobayashi S and Hachiya I.
The Journal of Organic Chemistry, 59(13), 3590-3596 (1994)
Opas Bunkoed et al.
Analytica chimica acta, 659(1-2), 251-257 (2010-01-28)
We report the development of transparent sol-gels with entrapped sensitive and selective reagents for the detection of formaldehyde. The sampling method is based on the adsorption of formaldehyde from the air and reaction with beta-diketones (for example acetylacetone) in a
Cindy S Ma et al.
The Journal of experimental medicine, 205(7), 1551-1557 (2008-07-02)
Hyper-immunoglobulin E syndrome (HIES) is a primary immune deficiency characterized by abnormal and devastating susceptibility to a narrow spectrum of infections, most commonly Staphylococcus aureus and Candida albicans. Recent investigations have identified mutations in STAT3 in the majority of HIES

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service