476404
2-Chloropentane
95%
Synonym(s):
1-Methylbutyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
95%
refractive index
n20/D 1.407 (lit.)
bp
94-95 °C/747 mmHg (lit.)
density
0.87 g/mL at 25 °C (lit.)
functional group
alkyl halide
chloro
SMILES string
CCCC(C)Cl
InChI
1S/C5H11Cl/c1-3-4-5(2)6/h5H,3-4H2,1-2H3
InChI key
NFRKUDYZEVQXTE-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
33.8 °F - closed cup
Flash Point(C)
1 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Organic Chemistry, 358-358 (2004)
Alkylations and Competing Rearrangements in the Aluminum Chloride Catalyzed Reactions of Secondary Alkyl Chlorides with Arenes.
The Journal of Organic Chemistry, 41(4), 659-665 (1976)
Preparation and properties of 2-and 3-chloropentanes.
Journal of the American Chemical Society, 60(10), 2536-2538 (1938)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service