Skip to Content
Merck
All Photos(3)

Key Documents

238376

Sigma-Aldrich

1-Chloropentane

99%

Synonym(s):

Amyl chloride, Pentyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4Cl
CAS Number:
Molecular Weight:
106.59
Beilstein:
1696936
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

107-108 °C (lit.)

solubility

alcohol: miscible(lit.)
diethyl ether: miscible(lit.)
water: insoluble(lit.)

density

0.882 g/mL at 25 °C (lit.)

functional group

alkyl halide

SMILES string

CCCCCCl

InChI

1S/C5H11Cl/c1-2-3-4-5-6/h2-5H2,1H3

InChI key

SQCZQTSHSZLZIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chloropentane forms asymmetric and symmetric pairs on the surface of Si (100)-2×1. Reaction of living polystyrene with 1-chloropentane (small molecule-polymer reaction) has been studied.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masateru M Ito et al.
Nature, 570(7761), 363-367 (2019-06-21)
The formation of microscopic cavities and microfibrils at stress hotspots in polymers is typically undesirable and is a contributor to material failure. This type of stress crazing is accelerated by solvents that are typically weak enough not to dissolve the
Kinetic study on reactions between polymer chain-ends. Coupling reactions of living polystyrene with chloro-ended polystyrene.
Okamoto A, et al.
Eur. Polymer J., 18(6), 545-548 (1982)
K R Harikumar et al.
Chemical communications (Cambridge, England), 47(44), 12101-12103 (2011-10-19)
Chloropentane forms asymmetric ('A') and symmetric ('S') pairs on Si(100)-2×1, differing in the direction of curvature of one pentane tail. Surprisingly this renders the rate of thermal reaction of 'A' fifteen times greater than 'S' in chlorinating room-temperature silicon. Correspondingly
Maxwell Wei-Hao Li et al.
Journal of chromatography. A, 1620, 461002-461002 (2020-03-08)
Micro gas chromatography (µGC) is a technique developed for rapid, in situ analysis of volatile organic compounds (VOCs) for environmental protection, industrial monitoring, and toxicology. While reduced µGC size and power requirements allow for increased portability, the low moisture and
Astrid Maruti et al.
Journal of chromatography. A, 1551, 41-51 (2018-04-14)
A novel extraction technique is proposed in which the Multiple Headspace Extraction (MHE) approach is used in conjunction with Headspace Sorptive Extraction (HSSE) and Gas Chromatography-Mass Spectrometry (GC-MS) detection. The extraction method was developed to determine volatile compounds in macroalgae.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service