Skip to Content
Merck
All Photos(3)

Key Documents

A55209

Sigma-Aldrich

1-(2-Aminoethyl)piperazine

99%

Synonym(s):

2-Piperazinoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N3
CAS Number:
Molecular Weight:
129.20
Beilstein:
104363
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.4 (vs air)

Quality Level

vapor pressure

0.05 mmHg ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.5 (lit.)

bp

218-222 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

NCCN1CCNCC1

InChI

1S/C6H15N3/c7-1-4-9-5-2-8-3-6-9/h8H,1-7H2

InChI key

IMUDHTPIFIBORV-UHFFFAOYSA-N

Application

Utilized in a variety of reactions for studying corrosion inhibition, biological activity, and metal ligand effects on catalysis.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J. Chem. Soc. Perkin Trans. II, 939-939 (1992)
S M Sami et al.
Journal of medicinal chemistry, 36(6), 765-770 (1993-03-19)
A new class of antitumor agents, having structural analogy to amonafide, but differing by the addition of a fourth ring in the nucleus, was synthesized conveniently from anthracene. Compounds with a variety of substituents, containing a basic nitrogen atom and
Chao Lin et al.
Journal of controlled release : official journal of the Controlled Release Society, 116(2), 130-137 (2006-11-03)
A group of novel poly(amido amine) homo- and copolymers (PAAs) containing secondary and tertiary amine groups in their main chain and different structures in the bisacrylamide segments were synthesized and evaluated as non-viral gene delivery vectors. Among these, also the
T Abdülkadir Coban et al.
Bioorganic & medicinal chemistry, 17(16), 5791-5795 (2009-07-29)
Sildenafil citrate, a phosphodiesterase-5 (PDE5) inhibitor widely used for the treatment of erectile dysfunction was investigated for its interaction with the zinc-enzyme carbonic anhydrase (CA, EC 4.2.1.1), as it has in its molecule a piperazine moiety also found in some
Jijin Gu et al.
Acta biomaterialia, 57, 156-169 (2017-04-27)
The active targeting of gene carriers is a powerful strategy for improving tumour-specific delivery and therapy. Although numerous l-peptide ligands play significant roles in the active targeting of nanomedicine, retro-inverso d-peptides have been explored as targeting ligands due to their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service