Skip to Content
Merck
All Photos(3)

Key Documents

471461

Sigma-Aldrich

2-(Ethylamino)ethanol

≥98%

Synonym(s):

N-Ethylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NHCH2CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
635671
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<1 mmHg ( 20 °C)

Quality Level

Assay

≥98%

refractive index

n20/D 1.441 (lit.)

bp

169-170 °C (lit.)

mp

−90 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

SMILES string

CCNCCO

InChI

1S/C4H11NO/c1-2-5-3-4-6/h5-6H,2-4H2,1H3

InChI key

MIJDSYMOBYNHOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

172.4 °F

Flash Point(C)

78 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis [2-(ethylamino) ethanol-2N, O] bis (saccharinato-N) cadmium (II).
Yilmaz VT and Kazak C.
Acta Crystallographica Section E, Structure Reports Online, 63(12), m2947-m2947 (2007)
Minhui He et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(3), 341-347 (2018-03-13)
The ability to directly detect alkaline phosphatase (ALP) activity in undiluted serum samples is of great importance for clinical diagnosis. In this work, we report the use of the distinctive metal-to-ligand charge-transfer (MLCT) absorption properties of the Cu(BCA)2+ (BCA =
Surface tension of aqueous binary mixtures of 2-(methylamino) ethanol and 2-(ethylamino) ethanol and aqueous ternary mixtures of these amines with triethanolamine or N-methyldiethanolamine from (293.15 to 323.15) K.
Alvarez E, et al.
Journal of Chemical and Engineering Data, 53(1), 318-321 (2007)
Corrosion Inhibition of a Carbon Steel by 2-Ethylamino-Ethanol in Aerated 3% NaCI Solutions-Effect of pH.
BUI N and Dabosi F.
Corrosion (Houston, TX, United States), 35(9), 392-397 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service