Skip to Content
Merck
All Photos(2)

Key Documents

471488

Sigma-Aldrich

2-(Diisopropylamino)ethanol

≥99%

Synonym(s):

N,N-Diisopropylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NCH2CH2OH
CAS Number:
Molecular Weight:
145.24
Beilstein:
1697955
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99%

refractive index

n20/D 1.442 (lit.)

bp

187-192 °C (lit.)

density

0.826 g/mL at 25 °C (lit.)

SMILES string

CC(C)N(CCO)C(C)C

InChI

1S/C8H19NO/c1-7(2)9(5-6-10)8(3)4/h7-8,10H,5-6H2,1-4H3

InChI key

ZYWUVGFIXPNBDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brown HC, et al.
Brown HC, et al.
Organometallics, 18(7), 1305-1309 (1999)
Separation and identification of some chemical warfare degradation products using electrospray high resolution ion mobility spectrometry with mass selected detection.
Asbury GR, et al.
Analytica Chimica Acta, 400(2), 273-283 (2000)
A pH-sensitive polymeric nanovesicle based on biodegradable poly (ethylene glycol)-b-poly (2-(diisopropylamino) ethyl aspartate) as a MRI-visible drug delivery system.
Sun Q, et al.
Journal of Materials Chemistry, 21(39), 15316-15326 (2011)
C Wu et al.
Analytical chemistry, 71(1), 273-278 (1999-01-01)
A surface ionization (SI) source was designed and constructed for ion mobility spectrometry (IMS). Compared with a conventional (63)Ni source, the surface ionization source is as simple and reliable, has an extended dynamic response range, is more selective in response
Bilal N Sheikh et al.
Nature cell biology, 22(7), 828-841 (2020-06-17)
Mutations in chromatin-modifying complexes and metabolic enzymes commonly underlie complex human developmental syndromes affecting multiple organs. A major challenge is to determine how disease-causing genetic lesions cause deregulation of homeostasis in unique cell types. Here we show that neural-specific depletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service