Skip to Content
Merck
All Photos(1)

Key Documents

467847

Sigma-Aldrich

Copper(II) chloride dihydrate

≥99.95% trace metals basis

Synonym(s):

Cupric chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2 · 2H2O
CAS Number:
Molecular Weight:
170.48
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

Assay

≥99.95% trace metals basis

form

solid

reaction suitability

core: copper
reagent type: catalyst

impurities

≤0.01% insolubles
≤500 ppm total metallic impurities

mp

100 °C (dec.) (lit.)

anion traces

nitrate (NO3-): ≤0.015%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Fe: ≤0.005%
K: ≤0.01%
Na: ≤0.02%
Ni: ≤0.01%

SMILES string

Cl[Cu]Cl.[H]O[H].[H]O[H]

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chlorinating agent; oxidizing agent; Lewis acid; catalyst for conjugate additions and acylation.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient synthesis of alpha,beta-acetylenic ketones.
P Li et al.
The Journal of organic chemistry, 66(11), 4087-4090 (2001-05-26)
Laura Engel et al.
Sensors (Basel, Switzerland), 19(5) (2019-03-13)
A fast and sensitive method to monitor hydrogen sulfide (H₂S) in ambient air based on a visible color change of a printed disposable sensor has been developed. As gas-sensitive material, an immobilized copper(II) complex of the azo dye 1-(2-pyridylazo)-2-naphtol (H-PAN)
Wenfeng Jiang et al.
Science (New York, N.Y.), 368(6491), 642-648 (2020-04-11)
The structural complexity of composite biomaterials and biomineralized particles arises from the hierarchical ordering of inorganic building blocks over multiple scales. Although empirical observations of complex nanoassemblies are abundant, the physicochemical mechanisms leading to their geometrical complexity are still puzzling
Bin Wu et al.
Carbohydrate research, 351, 68-73 (2012-03-01)
Two new glucoside esters 1 and 2 were produced as stress metabolites in the fresh leaves of Portulaca oleracea, in response to abiotic stress elicitation by CuCl(2). A new sugar ester (3) and two known compounds (4 and 5) were
Iron(II)-catalyzed trifluoromethylation of potassium vinyltrifluoroborates.
Andrew T Parsons et al.
Angewandte Chemie (International ed. in English), 51(12), 2947-2950 (2012-02-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service