Skip to Content
Merck
All Photos(3)

Key Documents

307483

Sigma-Aldrich

Copper(II) chloride dihydrate

ACS reagent, ≥99.0%

Synonym(s):

Cupric chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2 · 2H2O
CAS Number:
Molecular Weight:
170.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

vapor density

>1 (vs air)

Assay

≥99.0%

form

crystalline powder
crystals or granules

reaction suitability

reagent type: catalyst
core: copper

impurities

≤0.01% insolubles

mp

100 °C (dec.) (lit.)

anion traces

nitrate (NO3-): ≤0.015%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Fe: ≤0.005%
K: ≤0.01%
Na: ≤0.02%
Ni: ≤0.01%

SMILES string

Cl[Cu]Cl.[H]O[H].[H]O[H]

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Copper(II) chloride dihydrate can be used as a catalyst:
  • For the conversion of dimedone into N-arylhydrazinecarbothioamide based indazole derivatives by reacting with phenylhydrazine, hydrazine hydrate, and substituted arylisothiocyanates.
  • In the three-component Domino reaction ofarylboronic acids, arylcyanamides, and amines to synthesize 2-aminobenzimidazoles in the presence of bipyridine ligand.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deprotection of t-butyldimethylsiloxy (TBDMS) protecting group with catalytic copper (II) chloride dihydrate.
TAN ZP, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 11(9), 753-756 (2000)
Interaction of hydrazine with copper (II) chloride in acidic solutions. Formation, spectral and magnetic properties, and structures of copper (II), copper (I), and mixed-valence species.
Brown DB, et al.
Inorganic Chemistry, 18(10), 2635-2641 (1979)
An X-ray refinement of the crystal structure of copper (II) chloride dihydrate.
Engberg ?.
Acta Chemica Scandinavica, 24(10), 6-6 (1970)
Regeneration of carbonyl compounds from semicarbazones by copper (II) chloride dihydrate.
Ram RN and Varsha K.
Tetrahedron Letters, 32(41), 5829-5832 (1991)
Mónica João Barros Amorim et al.
Environmental pollution (Barking, Essex : 1987), 164, 164-168 (2012-03-01)
Environmental effects of copper nanoparticles are little studied in terrestrial ecosystems. In the present article, the toxicity of copper nanoparticles (Cu-NP) on the enchytraeid Enchytraeus albidus is compared to the toxicity of a copper-salt (CuCl(2)). The effect parameters studied were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service