Skip to Content
Merck
All Photos(3)

Key Documents

461121

Sigma-Aldrich

p-Toluidine

99%

Synonym(s):

4-Aminotoluene, 4-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
471281
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.9 (vs air)

Quality Level

vapor pressure

1 hPa ( 42 °C)
7 hPa ( 68 °C)

Assay

99%

form

powder, crystals or chunks

autoignition temp.

899 °F

expl. lim.

6.6 %

bp

200 °C (lit.)

mp

41-46 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N)cc1

InChI

1S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5H,8H2,1H3

InChI key

RZXMPPFPUUCRFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Toluidine is an aromatic amine. It can be detected in the water samples by homogeneous liquid-liquid extraction followed by ion mobility spectrometry (HLLE-IMS).

Application

p-Toluidine may be used for the determination of hemoglobin adducts of aromatic amines in nonsmokers and smokers of blond- or black-tobacco cigarettes. It may be used for the preparation of bidentate Schiff base ligand, via condensation with salicylaldehyde.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Sens. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amin Ashori et al.
Bulletin of environmental contamination and toxicology, 94(4), 474-478 (2014-11-28)
In this research, homogeneous liquid-liquid extraction followed by ion mobility spectrometry (HLLE-IMS) with corona discharge ionization source has been developed for the determination of p-toluidine. The analyte was extracted by single-phase extraction in a ternary solvent system and then the
M S Bryant et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(24), 9788-9791 (1988-12-01)
Hemoglobin adducts of 15 aromatic amines were determined in nonsmokers and smokers of blond- or black-tobacco cigarettes living in Turin, Italy. The subjects were all males age 55 or less and were representative of the population previously examined in a
Ruthenium (II) complexes containing bidentate Schiff bases and their antifungal activity.
Dharmaraj N, et al.
Transition Met. Chem. (London), 26(1-2), 105-109 (2001)
Paul G Stevenson et al.
Journal of chromatography. A, 1218(45), 8255-8263 (2011-10-11)
Several simple techniques are presented for the identification of the boundaries of chromatographic peaks. These methods provide a significant reduction in the time needed to perform the rapid, automatic calculation of the central peak moments and to evaluate the quality
Shunji Ito et al.
The Journal of organic chemistry, 70(6), 2285-2293 (2005-03-12)
[reaction: see text] N,N-Di(6-azulenyl)-p-toluidine (1a) and N,N,N',N'-tetra(6-azulenyl)-p-phenylenediamine (2a) and their derivatives with 1,3-bis(ethoxycarbonyl) substituents on each 6-azulenyl group (1b and 2b) were prepared by Pd-catalyzed amine azulenylation and characterized as a study into new aromatic amines for multistage amphoteric redox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service