Skip to Content
Merck
All Photos(1)

Key Documents

453757

Sigma-Aldrich

3-Cyclohexene-1-carboxylic acid

97%

Synonym(s):

1,2,3,6-Tetrahydrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9CO2H
CAS Number:
Molecular Weight:
126.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.48 (lit.)

bp

130-133 °C/4 mmHg (lit.)

mp

17 °C (lit.)

density

1.081 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C1CCC=CC1

InChI

1S/C7H10O2/c8-7(9)6-4-2-1-3-5-6/h1-2,6H,3-5H2,(H,8,9)

InChI key

VUSWCWPCANWBFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromination of 3-cyclohexene-1-carboxylic acid, epoxydation of methyl 3-cyclohexene-1-carboxylate and opening of methyl cis-and trans-3, 4-epoxycyclohexane-1-carboxylate: Stereochemical results.
Bellucci G, et al.
Tetrahedron, 28(13), 3393-3399 (1972)
Yi Yang et al.
Environmental science & technology, 48(4), 2344-2351 (2014-02-01)
The effect of halides on organic contaminant destruction efficiency was compared for UV/H2O2 and UV/S2O8(2-) AOP treatments of saline waters; benzoic acid, 3-cyclohexene-1-carboxylic acid, and cyclohexanecarboxylic acid were used as models for aromatic, alkene, and alkane constituents of naphthenic acids
Yeon Hee Ban et al.
Molecular bioSystems, 9(5), 944-947 (2012-12-12)
A FK506 analogue containing a non-natural starter unit was obtained through mutasynthesis by feeding cultures of Streptomyces sp. KCTC 11604BP fkbO deletion mutant with 3-cyclohexene-1-carboxylic acid. The structure of the new compound, 32-dehydroxy-FK506, and its biological activities were determined.
Karine Barral et al.
Journal of medicinal chemistry, 48(2), 450-456 (2005-01-22)
Starting from commercially available (rac)-3-cyclohexene-1-carboxylic acid, a series of purine and pyrimidine cis-substituted cyclohexenyl and cyclohexanyl nucleosides were synthesized through a key Mitsunobu reaction. Antiviral evaluations were performed on HIV, coxsackie B3, and herpes viruses (HSV-1, HSV-2, VZV, HCMV). Three
E R Blakley et al.
Canadian journal of microbiology, 28(12), 1324-1329 (1982-12-01)
A strain of Pseudomonas putida grew rapidly on cyclohexanecarboxylic acid as a sole source of carbon. A CoA-mediated beta-oxidation pathway was induced for the metabolism of the compound. The organism could not utilize 3-cyclohexenecarboxylic acid as a sole source of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service