Skip to Content
Merck
All Photos(3)

Key Documents

328324

Sigma-Aldrich

1-Cyclopentenecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H7CO2H
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

121-124 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)C1=CCCC1

InChI

1S/C6H8O2/c7-6(8)5-3-1-2-4-5/h3H,1-2,4H2,(H,7,8)

InChI key

PYRZPBDTPRQYKG-UHFFFAOYSA-N

Related Categories

General description

1-Cyclopentenecarboxylic acid was evaluated as a new effective anticonvulsant during Anticonvulsant Screening Program (ASP) of Antiepileptic Drug Development Program.

Application

1-Cyclopentenecarboxylic acid was used in synthesis of cis-8-hexahydroindanecarboxylic acid via Diels-Alder reaction with butadiene. It was also used in synthesis of isoxazole derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengcheng P Shao et al.
Bioorganic & medicinal chemistry letters, 19(18), 5334-5338 (2009-08-18)
A series of novel isoxazole voltage gated sodium channel blockers have been synthesized and evaluated. Substitutions on the benzylic position of benzamide were investigated to determine their effect on Na(v)1.7 inhibitory potency. The spirocyclobutyl substitution had the most significant enhancement
Marzanna Strupińska et al.
Acta poloniae pharmaceutica, 66(2), 155-159 (2009-09-02)
Previously obtained picolinic acid benzylamide is a potent anticonvulsant with low neurotoxicity. In search for new effective anticonvulsants twelve new benzylamides (1-12) were synthesized and preliminary evaluated in the Anticonvulsant Screening Program (ASP) of Antiepileptic Drug Development Program (ADDP) of
Preparation of cis-Hexahydroindane-8-carboxylic Acid.
DAUBEN W, et al.
The Journal of Organic Chemistry, 26(2), 297-300 (1961)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service