Skip to Content
Merck
All Photos(3)

Key Documents

101834

Sigma-Aldrich

Cyclohexanecarboxylic acid

98%

Synonym(s):

Hexahydrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CO2H
CAS Number:
Molecular Weight:
128.17
Beilstein:
970529
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

refractive index

n20/D 1.461 (lit.)

bp

232-233 °C (lit.)

mp

29-31 °C (lit.)

solubility

H2O: soluble 0.201g in 100g at 15 °C
organic solvents: soluble

density

1.033 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)C1CCCCC1

InChI

1S/C7H12O2/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2,(H,8,9)

InChI key

NZNMSOFKMUBTKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclohexanecarboxylic acid was used in a study to determine complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives[1].

Biochem/physiol Actions

Cyclohexanecarboxylic acid undergoes microbial degradation by a strain of Antherobacter to form para-hydroxybenzoic acid[2]. Cyclohexanecarboxylic acid undergoes aromatization and converts to Hippuric acid in rat liver extracts in vitro[3]. Cyclohexanecarboxylic acid is the starting reagent for the synthesis of polyketide-type antibiotics, Phoslactomycins[4].

Preparation Note

0.201g of cyclohexanecarboxylic acid dissolves in 100g water at 15°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Gadre et al.
Journal of pharmaceutical sciences, 86(2), 236-243 (1997-02-01)
Complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives (all possessing the carboxylic acid group) and with the series C6H5(CH2)nCOOH (n = 0 to 4) were measured in aqueous solution at 25 degrees C by potentiometry and
Biosynthesis of phoslactomycins: cyclohexanecarboxylic acid as the starter unit.
Sekiyama Y, et al.
Tetrahedron, 59(38), 7465-7471 (2003)
The microbial degradation of cyclohexanecarboxylic acid: a pathway involving aromatization to form p-hydroxybenzoic acid.
Blakley ER.
Canadian Journal of Microbiology, 20(10), 1297-1306 (1974)
Aromatization of cyclohexanecarboxylic acid.
B M Babior et al.
The Journal of biological chemistry, 241(16), 3643-3651 (1966-08-25)
Janice C Paslawski et al.
Biodegradation, 20(1), 125-133 (2008-07-18)
Naphthenic acids are a complex mixture of organic compounds which naturally occur in crude oil. Low molecular weight components of the naphthenic acids are known to be toxic in aquatic environments and there is a need to better understand the

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service