Skip to Content
Merck
All Photos(1)

Key Documents

448796

Sigma-Aldrich

Diethyl benzoylphosphonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COP(O)(OC2H5)2
CAS Number:
Molecular Weight:
242.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.508 (lit.)

bp

186 °C (lit.)

density

1.156 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(OCC)C(=O)c1ccccc1

InChI

1S/C11H15O4P/c1-3-14-16(13,15-4-2)11(12)10-8-6-5-7-9-10/h5-9H,3-4H2,1-2H3

InChI key

TZAMQIAPGYOUKF-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of polymeric metal complexes formed by reaction of excess diethyl acetyl-or diethyl benzoyl-phosphonate with metal chlorides at elevated temperatures.
Mikulski CM, et al.
J. Inorg. Nucl. Chem., 43(11), 2753-2757 (1981)
Studies on pyrimidine derivatives and related compounds. XLI. Reaction of diethyl benzoylphosphonate with thiamine (Takamizawa reaction 3).
A Takamizawa et al.
The Journal of organic chemistry, 31(9), 2951-2956 (1966-09-01)
Asymmetric Hydrogenation of a, ?-Unsaturated Ester-Phosphonates.
Huang Y, et al.
Advanced Synthesis & Catalysis, 351(9), 1423-1430 (2009)
Diethyl benzoylphosphonate as a ligand.
Mikulski CM, et al.
Transition Met. Chem. (London), 2(1), 135-140 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service