Skip to Content
Merck
All Photos(1)

Key Documents

T61301

Sigma-Aldrich

Triethyl phosphonoacetate

98%

Synonym(s):

(Diethoxyphosphinyl)acetic acid ethyl ester, Diethyl ethoxycarbonylmethylphosphonate, NSC 13898, NSC 16128, Triethyl carboxymethylphosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH2CO2C2H5
CAS Number:
Molecular Weight:
224.19
Beilstein:
1343714
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.431 (lit.)

bp

142-145 °C/9 mmHg (lit.)

density

1.13 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CP(=O)(OCC)OCC

InChI

1S/C8H17O5P/c1-4-11-8(9)7-14(10,12-5-2)13-6-3/h4-7H2,1-3H3

InChI key

GGUBFICZYGKNTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Horner-Wadsworth-Emmons reactions
  • Intramolecular Heck-type cyclization and isomerizations
  • Tsuji-Trost type reactions
  • Intramolecular aryne-ene reactions
  • Synthesis of aldehydes
Horner-Emmons-Wadsworth reagent recently used in the synthesis of chiral 2-methylcyclopropanecarboxylic acid from (S)-propylene oxide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Process Research & Development, 11, 689-689 (2007)
M Rodriguez et al.
International journal of peptide and protein research, 39(3), 273-277 (1992-03-01)
Racemic "carba" pseudo-dipeptide units such as Gly-psi(CH2-CH2)-D,L-Xaa were obtained either through the Horner-Emmons condensation of N-tert.-butyloxycarbonyl-beta alaninal with the appropriate substituted triethyl phosphonacetate, or from commercially available 3-carbethoxy-2-piperidone.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service