Skip to Content
Merck
All Photos(2)

Key Documents

427179

Sigma-Aldrich

Poly(vinylidene fluoride-co-hexafluoropropylene)

average Mw ~455,000, average Mn ~110,000, pellets

Synonym(s):

PVDF-HFP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(-CH2CF2-)x[-CF2CF(CF3)-]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

Quality Level

melt index

3.5-7.5 g/10 min (230°C/12.5kg)

mol wt

average Mn ~110,000
average Mw ~455,000

impact strength

6-10 ft-lb/in. (Izod, ASTM D 256, notched)

dielectric constant

9.0-10.0, 100 Hz (ASTM D 150)

hardness

70 (Shore D, ASTM D 2240)

refractive index

n20/D 1.42

viscosity

23,000-27,000 poise(230 °C) (100 sec-1)(lit.)

transition temp

Tm 155-160 °C (ASTM D 3418)

density

1.77 g/mL at 25 °C

SMILES string

FC(F)=C.FC(F)=C(F)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Application

Wire and cable coatings, flexible and corrosion-resistant tubing, and liners for pipes and tanks.

Features and Benefits

Excellent thermal stability, chemical and abrasion resistance, impervious to UV degradation, self-extinguishing and retains properties on aging. Improved flexibility at subzero temperatures, stress crack resistance and elongation to break over poly(vinylidene fluoride).

Physical form

Crystalline, thermoplastic copolymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service