Skip to Content
Merck
All Photos(1)

Key Documents

438340

Sigma-Aldrich

Trimethoxy(2-phenylethyl)silane

98%

Synonym(s):

(2-Phenylethyl)trimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2Si(OCH3)3
CAS Number:
Molecular Weight:
226.34
Beilstein:
3031224
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

95-96 °C/2 mmHg (lit.)

density

1.033 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCc1ccccc1)(OC)OC

InChI

1S/C11H18O3Si/c1-12-15(13-2,14-3)10-9-11-7-5-4-6-8-11/h4-8H,9-10H2,1-3H3

InChI key

UBMUZYGBAGFCDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethoxy(2-phenylethyl)silane (TMPES) is a phenyl functionalized trialkoxy silane that can be used as a siloxane based precursor.

Application

TMPES can be used as a silane-based coating on quantum dot (QD) light emitting diodes to improve the adhesion of QDs and improve the thermal stability of the device. It may be used in the synthesis of pure silica zeolite molecular sieves.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrolysis-condensation kinetics of different silane coupling agents
Brochier S, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 186(2), 240-254 (2011)
Quantum dot-layer-encapsulated and phenyl-functionalized silica spheres for highly luminous, colour rendering, and stable white light-emitting diodes
Yoo H, et al.
Nanoscale, 7(30), 12860-12867 (2015)
Organic-functionalized molecular sieves as shape-selective catalysts
Jones CW, et al.
Nature, 393(6680), 52-52 (1998)
Brandy J Johnson et al.
Sensors (Basel, Switzerland), 11(1), 886-904 (2012-02-22)
The development of porphyrin-embedded mesoporous organosilicate materials for application to the detection of volatile hydrocarbon solvents is described. Design of the receptor and optical indicator construct begins with parallel selection of the porphyrin indicator and design of the mesoporous sorbent.
Louise Stjern et al.
International journal of pharmaceutics, 531(2), 595-605 (2017-05-31)
Cyclodextrins (CDs) and mesoporous silica particles (MSPs) have been combined as composite carriers for controlled antibiotic release. CDs were employed as "gatekeeper" agents and grafted onto MSPs to retain drug molecules inside the MSP carrier. A variety of CDs (unfunctionalized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service