Skip to Content
Merck
Trimethoxy(2-phenylethyl)silane 98%

Trimethoxy(2-phenylethyl)silane

Trimethoxy(2-phenylethyl)silane 98%
Synonym(s):
(2-Phenylethyl)trimethoxysilane
Linear Formula:
C6H5CH2CH2Si(OCH3)3
CAS Number:
Molecular Weight:
226.34
EC Number:

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing