All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H8N4O2
CAS Number:
Molecular Weight:
156.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
123-127 °C (lit.)
functional group
ester
SMILES string
CCOC(=O)Cc1nnn[nH]1
InChI
1S/C5H8N4O2/c1-2-11-5(10)3-4-6-8-9-7-4/h2-3H2,1H3,(H,6,7,8,9)
InChI key
NAOHMNNTUFFTBF-UHFFFAOYSA-N
General description
Ethyl 1H-tetrazole-5-acetate is a tetrazole derivative. Its effect on glutaminyl cyclase activity of the recombinant human glutaminyl cyclase (QC) has been reported.
Application
Ethyl 1H-tetrazole-5-acetate (Ethyl 1H-tetrazole-5-yl acetate, Hetza) may be employed as starting reagent for the synthesis of ethyl aryloxadiazolylacetates. It may be used for the preparation of homochiral 3D diamondoid metal-organic framework [Zn2(etza)4].
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of 5-aryl-1, 3, 4-oxadiazolyl-2-acetic acids.
Janda L.
Heterocyclic Communications, 7(5), 411-416 (2001)
A spontaneously resoluted zinc-organic framework with nonlinear optical and ferroelectric properties generated from tetrazolate-ethyl ester ligand.
Liang X-Q, et al.
CrystEngComm, 12(11), 3499-3501 (2010)
Kai-Fa Huang et al.
Protein expression and purification, 43(1), 65-72 (2005-08-09)
Glutaminyl cyclase (QC) catalyzes the N-terminal pyroglutamate formation of numerous hormones and peptides from their glutaminyl precursor. Pyroglutamate is a posttranslational or cotranslational modification important in many physiological and pathological processes. Here, we present the cloning of a QC cDNA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service