Skip to Content
Merck
All Photos(1)

Key Documents

415308

Sigma-Aldrich

1-Ethylpiperazine

98%

Synonym(s):

N-Ethylpiperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
Beilstein:
102971
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.469 (lit.)

bp

157 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

SMILES string

CCN1CCNCC1

InChI

1S/C6H14N2/c1-2-8-5-3-7-4-6-8/h7H,2-6H2,1H3

InChI key

WGCYRFWNGRMRJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Ethylpiperazine may be used in the preparation of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amjad M Qandil
Pharmaceuticals (Basel, Switzerland), 5(5), 460-468 (2012-01-01)
A group of benzimidazole analogs of sildenafil, 3-benzimidazolyl-4-methoxy-phenylsulfonylpiperazines 2-4 and 3-benzimidazolyl-4-methoxy-N,N-dimethyl- benzenesulfonamide (5), were efficiently synthesized. Compounds 2-5 were characterized by NMR and MS and contrary to the reported mass spectra of sildenafil, the spectra of the piperazine-containing compounds 2-4
pKa Values of Some Piperazines at (298, 303, 313, and 323)K.
Khalili F, et al.
Journal of Chemical and Engineering Data, 54(10), 2914-2917 (2009)
Surajit Maity et al.
Physical chemistry chemical physics : PCCP, 12(23), 6150-6156 (2010-04-14)
The structures of the binary complexes between phenylacetylene and several tertiary amines viz., triethylamine, 1-ethylpiperidine, 1-ethylpiperazine, 1-azabicyclo[2.2.2]octane, and 1,4-diazabicyclo[2.2.2]octane were inferred using infrared-optical double resonance spectroscopy. The IR spectra in the acetylenic C-H stretching region clearly rule out the formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service