Skip to Content
Merck
All Photos(1)

Key Documents

511617

Sigma-Aldrich

6-Chloropurine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3ClN4
CAS Number:
Molecular Weight:
154.56
Beilstein:
5774
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

>300 °C (dec.) (lit.)

SMILES string

Clc1ncnc2[nH]cnc12

InChI

1S/C5H3ClN4/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H,7,8,9,10)

InChI key

ZKBQDFAWXLTYKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Chloropurine (6-CIPH), a 6-substituted purine derivative, is an antileukemic drug. It can be prepared by the chlorination of hypoxanthine with phosphorus oxychloride in the presence of dimethylaniline. The NMR-based conformational analysis of the products formed during the reaction of 6-CIPH with 3,4-di-O-acetyl-D-xylal and 3,4-di-O-acetyl-L-arabinal have been reported. 6-CIPH can undergo palladium-catalyzed cross coupling with organostannanes at 6-position to form the corresponding arylated or alkylated products.

Application

6-Chloropurine may be used:
  • To prepare purine via catalytic dehydrogenation.
  • To prepare 9-alkylated adenines via Mitsunobu reaction with various alcohols.
  • As a starting material to synthesize dihydroisoxazole 6-chloropurine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis and anti-HIV activity of dihydroisoxazole 6-chloropurine and adenine.
Xiang Y, et al.
Bioorganic & Medicinal Chemistry Letters, 6(9), 1051-1054 (1996)
The Synthesis and Properties of 6-Chloropurine and Purine1.
Bendich A, et al.
Journal of the American Chemical Society, 76(23), 6073-6077 (1954)
REACTIONS OF RIBONUCLEOTIDE DERIVATIVES OF PURINE ANALOGUES AT THE CATALYTIC SITE OF INOSINE 5'-PHOSPHATE DEHYDROGENASE.
A HAMPTON
The Journal of biological chemistry, 238, 3068-3074 (1963-09-01)
6-Chloropurines and organostannanes in palladium catalyzed cross coupling reactions.
Gundersen LL.
Tetrahedron Letters, 35(19), 3155-3158 (1994)
Synthesis of Nucleosides and Related Compounds. Part XXV. The Alkylation of 6-Chloropurine with Alcohols by Mitsunobu Reaction.
Toyota A, et al.
Chemical & Pharmaceutical Bulletin, 40(4), 1039-1041 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service