Skip to Content
Merck
All Photos(1)

Key Documents

413550

Sigma-Aldrich

Bisphenol A ethoxylate diacrylate

average Mn ~468, EO/phenol 1.5, contains 250 ppm MEHQ as inhibitor

Synonym(s):

Bisphenol A polyethylene glycol diether diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHCO2(CH2CH2O)nC6H4-4-]2C(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

mol wt

average Mn ~468

contains

250 ppm MEHQ as inhibitor

composition

EO/phenol, 1.5

refractive index

n20/D 1.545

density

1.146 g/mL at 25 °C

SMILES string

OCCO.OC(=O)C=C.CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

Looking for similar products? Visit Product Comparison Guide

General description

Bisphenol A ethoxylate diacrylate is a cross-linking monomer that is used to improve the mechanical properties of the polymeric system. It is majorly used in the formation of solid polymer based electrolytes which can be used for electrochemical applications.

Application


  • Removable acrylic pressure-sensitive adhesives activated by UV-radiation: The preparation of bisphenol A ethoxylate diacrylate through the condensation reaction of BADGE and AA is detailed for adhesive applications (Chu et al., 2014).

  • Thermomechanical Properties and Shape‐Memory Behavior of Bisphenol A Diacrylate‐Based Shape‐Memory Polymers: The study analyzes the shape-memory properties of polymers based on bisphenol A diacrylate and BAEDA (Santiago et al., 2016).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A study of cross-linked PEO gel polymer electrolytes using bisphenol A ethoxylate diacrylate: ionic conductivity and mechanical properties
Kang Y, et al.
Journal of Power Sources, 119(3), 432-437 (2003)
Synthesis of clay nanocomposite materials by light-induced crosslinking polymerization
Benfarhi S, et al.
European Polymer Journal, 40(3), 493-501 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service