416525
Bisphenol A ethoxylate
average Mn ~492, EO/phenol 3
Synonym(s):
2,2-Bis(4-hydroxyphenyl)propane ethylene oxide adduct, Bisphenol A-ethylene oxide adduct, Ethoxylated bisphenol A
About This Item
Recommended Products
form
viscous liquid
Quality Level
mol wt
average Mn ~492
composition
EO/phenol, 3
refractive index
n20/D 1.542
density
1.14 g/mL at 25 °C
SMILES string
OCCO.CC(C)(c1ccc(O)cc1)c2ccc(O)cc2
InChI
1S/C15H16O2.C2H6O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12;3-1-2-4/h3-10,16-17H,1-2H3;3-4H,1-2H2
InChI key
PFZPRMFEGRUXTO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service