Skip to Content
Merck
All Photos(3)

Key Documents

413348

Sigma-Aldrich

N1-(3-Trimethoxysilylpropyl)diethylenetriamine

technical grade

Synonym(s):

2-[2-(3-Trimethoxysilylpropylamino)ethylamino]ethylamine, 3-[2-(2-Aminoethylamino)ethylamino]propyltrimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3Si(CH2)3NHCH2CH2NHCH2CH2NH2
CAS Number:
Molecular Weight:
265.43
Beilstein:
8421470
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor pressure

1 mmHg ( 20 °C)

form

liquid

refractive index

n20/D 1.459 (lit.)

bp

114-118 °C/2 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CO[Si](CCCNCCNCCN)(OC)OC

InChI

1S/C10H27N3O3Si/c1-14-17(15-2,16-3)10-4-6-12-8-9-13-7-5-11/h12-13H,4-11H2,1-3H3

InChI key

NHBRUUFBSBSTHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N1-(3-Trimethoxysilylpropyl)diethylenetriamine (TRI-silane) can be used as a grafting agent to incorporate amine functional group onto the surface of various ordered mesoporous silica. It can be also used in CO2 capture applications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

JinHyeong Lee et al.
Scientific reports, 10(1), 4882-4882 (2020-03-19)
In this study, the influence of drying conditions on amine (-NH3) functionalization of graphene oxide (GO) was evaluated, and the hexavalent chromium (Cr(VI)) adsorption efficiency of the prepared materials was compared. 3-[2-(2-aminoehtylamino) ethylamino]propyl-trimethoxysilane (3N) was used for amine functionalization. The
N 1-(3-Trimethoxysilylpropyl) diethylenetriamine grafted KIT-6 for CO2/N2 selective separation
Kishor, Rupak and Ghoshal, Aloke Kumar
Royal Society of Chemistry Advances, 6(2), 898-909 (2016)
Modeling adsorption of CO2 on amine-functionalized mesoporous silica. 2: Kinetics and breakthrough curves
S-G Rodrigo and Sayari A
Chemical Engineering Journal, 161(1-2), 182-190 (2010)
Modeling CO2 adsorption on amine-functionalized mesoporous silica: 1. A semi-empirical equilibrium model
S-G, Rodrigo, et al.
Chemical Engineering Journal, 161(1-2), 173-181 (2010)
Takahito Nakagawa et al.
Journal of biotechnology, 116(2), 105-111 (2005-01-25)
A simple microchip device for DNA extraction was constructed based on electrostatic interactions between surface amine groups and DNA. Microchannel was fabricated on silicon wafer by photolithography and coated with 3-aminopropyltriethoxysilane (APTES) or 3-[2-(2-aminoethylamino)-ethylamino]-propyltrimethoxysilane (AEEA) to introduce amine groups on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service