Skip to Content
Merck
All Photos(3)

Key Documents

A3648

Sigma-Aldrich

(3-Aminopropyl)triethoxysilane

≥98%

Synonym(s):

APTS, 3-Triethoxysilylpropylamine, APTES

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)3Si(OC2H5)3
CAS Number:
Molecular Weight:
221.37
Beilstein:
1754988
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥98%

form

liquid

pH

11 (20 °C, 20 g/L)

bp

217 °C/760 mmHg (lit.)

mp

220-222 °C

density

0.946 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCO[Si](CCCN)(OCC)OCC

InChI

1S/C9H23NO3Si/c1-4-11-14(12-5-2,13-6-3)9-7-8-10/h4-10H2,1-3H3

InChI key

WYTZZXDRDKSJID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • (3-Aminopropyl)triethoxysilane has been used for the preparation of cadherin surfaces on glass slides.
  • It has been used for the preparation of RGD (Arg-Gly-Asp)-treated silicone membranes.
  • It has been used for the preparation of the avidin–biotin coverslips.

Biochem/physiol Actions

(3-Aminopropyl)triethoxysilane forms aminopropyl derivative of glass, an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Apoptosis and survival of osteoblast-like cells are regulated by surface attachment.
Grigoriou V, et al.
The Journal of Biological Chemistry, 280, 1733-1739 (2005)
Real-time observation of a single DNA digestion by lambda exonuclease under a fluorescence microscope field.
Matsuura S, et al.
Nucleic Acids Research, 29, E79-E79 (2001)
Vladimir Gubala et al.
Analytica chimica acta, 760, 75-82 (2012-12-26)
Herein a simple analytical method is presented for the characterization of biomolecule adsorption on cyclo olefin polymer (COP, trade name: Zeonor(®)) substrates which are widely used in microfluidic lab-on-a-chip devices. These Zeonor(®) substrates do not possess native functional groups for
Isabelle Bonnet et al.
Nucleic acids research, 36(12), 4118-4127 (2008-06-12)
The restriction endonuclease EcoRV can rapidly locate a short recognition site within long non-cognate DNA using 'facilitated diffusion'. This process has long been attributed to a sliding mechanism, in which the enzyme first binds to the DNA via nonspecific interaction
Sri Ram Krishna Vedula et al.
Nature communications, 6, 6111-6111 (2015-01-23)
The closure of gaps within epithelia is crucial to maintain its integrity during biological processes such as wound healing and gastrulation. Depending on the distribution of extracellular matrix, gap closure occurs through assembly of multicellular actin-based contractile cables or protrusive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service