Skip to Content
Merck
All Photos(3)

Key Documents

399558

Sigma-Aldrich

Lithium nitride

greener alternative

−60 mesh, ≥99.5%

Synonym(s):

Trilithium nitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li3N
CAS Number:
Molecular Weight:
34.83
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.5%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

−60 mesh

mp

840-850 °C (N2)

density

1.3 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li]N([Li])[Li]

InChI

1S/3Li.N

InChI key

IDBFBDSKYCUNPW-UHFFFAOYSA-N

General description

Lithium nitride (Li3N) is a solid-state ionic conductor with an ionic conductivity of ~6 x 10−3 Scm−1 and with a high Young′s modulus. It can generally be prepared by placing Li metal under nitrogen environment.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Li3N can be used in a variety of applications such as a hydrogen storage material, an electron injection material for organic light-emitting diodes (OLEDs), and in the fabrication of lithium-ion batteries.

Features and Benefits

Forms high room-temperature conductivity, high lithium conduction materials with SiS2 with excellent solid electrolyte properties.

Analysis Note

Hydrogen content, XRD plots and metal purity data are available upon request.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermally decomposable lithium nitride as an electron injection material for highly efficient and stable OLEDs
Duan L, et al.
The Journal of Physical Chemistry C, 113(30), 13386-13390 (2009)
Interaction of nitrogen with lithium in lithium ion batteries
Wang H, et al.
Solid State Ionics, 180(2-3), 212-215 (2009)
Joshua W Makepeace et al.
Physical chemistry chemical physics : PCCP, 20(35), 22689-22697 (2018-08-24)
Lithium imide is a promising new catalyst for the production of hydrogen from ammonia. Its catalytic activity has been reported to be significantly enhanced through its use as a composite with various transition metal nitrides. In this work, two of
D Muller-Bouvet et al.
Physical chemistry chemical physics : PCCP, 19(40), 27204-27211 (2017-10-04)
A large data set of XAS (X-ray Absorption Spectroscopy) Manganese K-edge spectra has been collected operando and studied upon the electrochemical oxidation of the promising Li-ion battery anode material Li
Boitumelo J Matsoso et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-04)
Hexagonal boron nitrite (hBN) is an attractive material for many applications such as in electronics as a complement to graphene, in anti-oxidation coatings, light emitters, etc. However, the synthesis of high-quality hBN at cost-effective conditions is still a great challenge.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service