Skip to Content
Merck
All Photos(1)

Key Documents

398780

Sigma-Aldrich

2,4,6-Tribromoanisole

99%

Synonym(s):

2,4,6-Tribromo-1-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br3C6H2OCH3
CAS Number:
Molecular Weight:
344.83
Beilstein:
2210361
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

297-299 °C (lit.)

mp

84-88 °C (lit.)

functional group

bromo

SMILES string

COc1c(Br)cc(Br)cc1Br

InChI

1S/C7H5Br3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3H,1H3

InChI key

YXTRCOAFNXQTKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,6-Tribromoanisole has been reported to cause cork taint in wines and vortex assisted liquid-liquid microextraction (VALLME) method for its determination has been developed. It is a musty-smelling metabolite of fungicide 2,4,6-tribromophenol. Determination of 2,4,6-trichloroanisole in wine at low ngL-1 levels by gas chromatography-high-resolution mass spectrometry (GC-HRMS) method has been repoted.

Application

2,4,6-Tribromoanisole is suitable reagent used for the determination of 2,4,6-tribromoanisole in packaging materials and food by an analytical method based on multipleion detection GC-MS.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Jönsson et al.
Journal of chromatography. A, 1111(1), 71-75 (2006-02-09)
A gas chromatography-high-resolution mass spectrometry (GC-HRMS) method using solid-phase microextraction (SPME) for the determination of 2,4,6-trichloroanisole (TCA) and 2,4,6-tribromoanisole (TBA) in wine at low ng L(-1) levels was developed. A robust SPME method was developed by optimizing several different parameters
2, 4, 6-Tribromoanisole: a potential cause of mustiness in packaged food.
Whitfield FB, et al.
Journal of Agricultural and Food Chemistry, 45(3), 889-893 (1997)
C Pizarro et al.
Talanta, 128, 1-8 (2014-07-26)
A vortex assisted liquid-liquid microextraction (VALLME) method was developed and optimised for the determination of the main compounds that can cause cork taint in wines, 2,4,6-trichloroanisole (TCA), 2,3,4,6-tetrachloroanisole (TeCA), 2,4,6-tribromoanisole (TBA) and pentachloroanisole (PCA); and their corresponding halophenolic precursors. Target
Navideh Sadoughi et al.
Journal of agricultural and food chemistry, 63(11), 2877-2885 (2015-02-24)
An optimized method for the quantitation of volatile compounds responsible for off-aromas, such as earthy odors, found in wine and grapes was developed. The method involved a fast and simple headspace solid-phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) for simultaneous determination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service