Skip to Content
Merck
All Photos(2)

Key Documents

137715

Sigma-Aldrich

2,4,6-Tribromophenol

99%

Synonym(s):

1,3,5-Tribromo-2-hydroxybenzene, Bromol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br3C6H2OH
CAS Number:
Molecular Weight:
330.80
Beilstein:
776920
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

11.4 (vs air)

Assay

99%

bp

282-290 °C/746 mmHg (lit.)

mp

90-94 °C (lit.)

solubility

water: soluble 0.007g/100ml at 25 °C
chloroform: soluble
diethyl ether: soluble
ethanol: soluble
methanol: soluble
methylene chloride: soluble

functional group

bromo

SMILES string

Oc1c(Br)cc(Br)cc1Br

InChI

1S/C6H3Br3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

BSWWXRFVMJHFBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,6-Tribromophenol undergoes oxidative degradation catalyzed by SiO2-supported iron(III)-5,10,15,20-tetrakis(4-carboxyphenyl) porphyrin. It is degraded by Bacillus sp.GZT strain via reductive bromination as major degradation pathway. It is a widely used brominated flame retardant.

Application

2,4,6-Tribromophenol was used in analysis of nonylphenol, nonylphenol monoethoxylate and octylphenol in sediment, water and fish tissue.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estrogenic alkylphenols in fish tissues, sediments, and waters from the UK Tyne and Tees estuaries.
Lye CM, et al.
Environmental Science & Technology, 33(7), 1009-1014 (1999)
Qianqian Zhu et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 48(13), 1593-1601 (2013-08-21)
Iron(III)-porphyrin complexes are generally regarded as green catalysts, since they mimic the catalytic center of cytochrome-P450 and widely used as green catalysts for degrading halogenated phenols in wastewater, such as landfill leachates. However, iron(III)-porphyrins are deactivated by self-oxidation in the
Taiki Futagami et al.
Applied and environmental microbiology, 75(21), 6905-6909 (2009-09-15)
Halogenated organic compounds serve as terminal electron acceptors for anaerobic respiration in a diverse range of microorganisms. Here, we report on the widespread distribution and diversity of reductive dehalogenase homologous (rdhA) genes in marine subsurface sediments. A total of 32
Natsuko Kajiwara et al.
Journal of hazardous materials, 192(3), 1250-1259 (2011-07-26)
The concentrations of traditional brominated flame retardants (BFRs) and organophosphate flame retardants (OPFRs) in new consumer products, including electronic equipment, curtains, wallpaper, and building materials, on the Japanese market in 2008 were investigated. Although some components of the electronic equipment
Hidetoshi Kuramochi et al.
Journal of environmental monitoring : JEM, 10(2), 206-210 (2008-02-05)
2,4,6-Tribromophenol (TBP) is expected to exist in both ionic and non-ionic forms in the environment due to ionisation of the phenolic group at near neutral pH. In this study, the water solubility (Sw) and 1-octanol-water partition coefficient (Kow) of aqueous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service