Skip to Content
Merck
All Photos(1)

Key Documents

392049

Sigma-Aldrich

1-Pyrenesulfonic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10O3S · xH2O
CAS Number:
Molecular Weight:
282.31 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

125-129 °C (lit.)

Quality Level

solubility

DMSO: soluble 250 mg/10 mL, clear to slightly hazy, yellow to brown

SMILES string

[H]O[H].OS(=O)(=O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C16H10O3S.H2O/c17-20(18,19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11;/h1-9H,(H,17,18,19);1H2

InChI key

RRDYFCSDPUHYDA-UHFFFAOYSA-N

Application

1-Pyrenesulfonic acid hydrate (PSA) can be used as a reactant to functionalize the surface of the graphene oxide along with tetrakis-(4-hydroxylphenyl)porphyrin (THPP). The synthesized graphene oxide nanohybrid (GO/THPP/PSA) material exhibited excellent photocatalytic activity for the production of hydrogen. It is also used as a building block to prepare pyrene-containing organostannoxanes by reacting with various organotin precursors.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication mechanism and photocatalytic activity for a novel graphene oxide hybrid functionalized with tetrakis-(4-hydroxylphenyl) porphyrin and 1-pyrenesulfonic acid
Luo Q, et al.
Applied Surface Science, 427, 15-23 (2018)
Pyrene sulfonate functionalised single-walled carbon nanotubes for mediatorless dioxygen bioelectrocatalysis.
JN Martin, et al.
Electrochemical Communications, 11(5), 1042-1044 (2009)
Keun-Hwan Oh et al.
ACS applied materials & interfaces, 6(10), 7751-7758 (2014-04-09)
We report boron nitride nanoflakes (BNNFs), for the first time, as a nanofiller for polymer electrolyte membranes in fuel cells. Utilizing the intrinsic mechanical strength of two-dimensional (2D) BN, addition of BNNFs even at a marginal content (0.3 wt %)
Subrata Kundu et al.
Chemistry, an Asian journal, 9(5), 1403-1412 (2014-03-13)
Organostannoxanes have been used as scaffolds for the preparation of multi-chromophore assemblies. A single-step synthesis procedure allows the preparation of compounds in which the number of chromophore units can be varied from two to six. Thus, the reactions of pyrene
Preparation of Free 1-Pyrenesulfonic Acid
Abe Y and Hsu PC.
Yuki Gosei Kaguku Kyokaishi, 33(4), 277-278 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service