Skip to Content
Merck
All Photos(1)

Key Documents

391581

Sigma-Aldrich

1-Pyrenecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H10O2
CAS Number:
Molecular Weight:
246.26
Beilstein:
2375854
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

mp

270-272 °C (lit.)

SMILES string

OC(=O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C17H10O2/c18-17(19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9H,(H,18,19)

InChI key

HYISVWRHTUCNCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Pyrenecarboxylic acid (PCA) is a polyaromatic derivative that has an amphiphilic characteristic. It is a fluorophore that shows absorption spectra in the UV region. PCA has a carboxylic acid group that attaches with pyrene nucleus. It can be used as a sensitizer that facilitates the electron transport from one medium to another.

Application

Functionalized pyrene for synthesis.
PCA can be used in the surface modification of graphene and carbon nanotubes (CNTs), which can further be used for a variety of electronic applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haodong Liu et al.
Advanced materials (Deerfield Beach, Fla.), 33(8), e2004782-e2004782 (2021-01-16)
The revolutionary and pioneering advancements of flexible electronics provide the boundless potential to become one of the leading trends in the exploitation of wearable devices and electronic skin. Working as substantial intermediates for the collection of external mechanical signals, flexible
Optical and sensing properties of 1-pyrenecarboxylic acid-functionalized graphene films laminated on polydimethylsiloxane membranes
An X, et al.
ACS Nano, 5(2), 1003-1011 (2011)
Noncovalent functionalization as an alternative to oxidative acid treatment of single wall carbon nanotubes with applications for polymer composites
Simmons TJ, et al.
ACS Nano, 3(4), 865-870 (2009)
Tunable graphene-silicon heterojunctions for ultrasensitive photodetection
An X, et al.
Nano Letters, 13(3), 909-916 (2013)
El Hadji Mamour Sakho et al.
Materials science & engineering. C, Materials for biological applications, 95, 43-48 (2018-12-24)
We herein report the synthesis of MFe2O4 (M = Mn, Mg)/reduced graphene oxide (MFe2O4/RGO) through a simple and novel pressure cooker assisted solvothermal method. The structure and morphology of the as-prepared materials were investigated using X-ray diffraction (XRD), transmission electron microscopic (TEM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service