Skip to Content
Merck
All Photos(1)

Key Documents

D201146

Sigma-Aldrich

Dioctylamine

97%

Synonym(s):

Di-n-Octylamine, N,N-Dioctylamine, N-Octyl-1-octanamine, N-n-Octyl-n-octylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7NH(CH2)7CH3
CAS Number:
Molecular Weight:
241.46
Beilstein:
1748376
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.3 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.4432 (lit.)

bp

297-298 °C (lit.)

mp

13-16 °C (lit.)

density

0.799 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCNCCCCCCCC

InChI

1S/C16H35N/c1-3-5-7-9-11-13-15-17-16-14-12-10-8-6-4-2/h17H,3-16H2,1-2H3

InChI key

LAWOZCWGWDVVSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dioctylamine can be used as a reactant in the synthesis of:
  • Ionic liquids based on the tetra-alkyl-dimethylguanidinium cation
  • N-(2,5-di-tert-butylphenyl)perylene-3,4-dicarboximide based perylene photosensitizers.

It can be also employed as a surfactant in the shape-controlled synthesis of some nanoparticles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New perylenes for dye sensitization of TiO2
Suzanne F and Brian AG
New. J. Chem., 26, 1155-1160 (2002)
Nanomagnetism and spin electronics: materials, microstructure and novel properties
Krishnan KM, et al.
Journal of materials science & technology, 41(3), 793-815 (2006)
Synthesis and properties of tetra-alkyl-dimethylguanidinium salts as a potential new generation of ionic liquids
Nuno MMM, et al.
Green Chemistry, 5, 347-352 (2003)
Crystal S Lewis et al.
Nanoscale, 8(4), 2129-2142 (2016-01-05)
We report on the synthesis and structural characterization of novel semiconducting heterostructures composed of cadmium selenide (CdSe) quantum dots (QDs) attached onto the surfaces of novel high-surface area, porous rare-earth-ion doped alkaline earth titanate micron-scale spherical motifs, i.e. both Eu-doped
Federico Bertasi et al.
Physical chemistry chemical physics : PCCP, 19(38), 26230-26239 (2017-09-22)
This work describes the preparation of the new lipophilic ionic liquid tetraoctyl-formamidinium bis(trifluoromethanesulfonyl) imide (TOFATFSI), which is miscible with lower alkanes. In particular, this work focuses on the electric behaviour of TOFATFSI in the particularly challenging highly apolar environment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service