Skip to Content
Merck
All Photos(1)

Key Documents

375705

Sigma-Aldrich

2-Methyl-3-(trifluoromethyl)aniline

95%

Synonym(s):

2-Amino-6-(trifluoromethyl)toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(CF3)NH2
CAS Number:
Molecular Weight:
175.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

38-42 °C (lit.)

functional group

fluoro

SMILES string

Cc1c(N)cccc1C(F)(F)F

InChI

1S/C8H8F3N/c1-5-6(8(9,10)11)3-2-4-7(5)12/h2-4H,12H2,1H3

InChI key

TWLDBACVSHADLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-3-(trifluoromethyl)aniline belongs to the trifluoromethylbenzene series.
Improved synthesis of 2-methyl-3-(trifluoromethyl)aniline has been reported.

Application

2-Methyl-3-(trifluoromethyl)aniline may be used in chemical synthesis.
2-Methyl-3-(trifluoromethyl)aniline may be used in the synthesis of an analgesic compound, flunixin, via reaction with 2-chloronicotinate in ethylene glycol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

205.0 °F - closed cup

Flash Point(C)

96.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved Process for the Preparation of 2 Methyl-3-trifluoromethylaniline: A Versatile Intermediate for Flunixin Synthesis.
Jaouhari R and Quinn P.
ChemInform, 26(3) (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service