Skip to Content
Merck
All Photos(1)

Key Documents

337056

Sigma-Aldrich

4-(Trifluoromethoxy)aniline

98%

Synonym(s):

α,α,α-Trifluoro-p-anisidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4NH2
CAS Number:
Molecular Weight:
177.12
Beilstein:
2090209
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.463 (lit.)

bp

73-75 °C/10 mmHg (lit.)

density

1.32 g/mL at 20 °C (lit.)

functional group

fluoro

SMILES string

Nc1ccc(OC(F)(F)F)cc1

InChI

1S/C7H6F3NO/c8-7(9,10)12-6-3-1-5(11)2-4-6/h1-4H,11H2

InChI key

XUJFOSLZQITUOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-(Trifluoromethoxy)aniline was used in the synthesis of:
  • side-group liquid-crystalline polymethacrylates with fluorine-containing mesogens
  • derivatives of 3-(quinolin-3-yl)acrylates
  • series of novel Shiff bases, via condensation with pyridinecarboxaldehydes in the presence of molecular sieves

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Addition of Me3SiCN to trifluoromethyl derivates of N-(pyridylmethylidene) anilines catalyzed by Lewis acids.
Iovel I, et al.
Applied Organometallic Chemistry, 15(9), 733-743 (2001)
Microwave-assisted multistep synthesis of functionalized 4-arylquinolin-2 (1 H)-ones using palladium-catalyzed cross-coupling chemistry.
Glasnov TN, et al.
The Journal of Organic Chemistry, 70(10), 3865-3870 (2005)
Synthesis of liquid-crystalline poly (meth) acrylates with 4-trifluoromethoxy-azobenzene mesogenic side-groups.
Prescher D, et al.
Journal of Fluorine Chemistry, 74(2), 185-189 (1995)
M Tugnait et al.
Journal of pharmaceutical and biomedical analysis, 28(5), 875-885 (2002-06-01)
A combination of 19F, 1H NMR and HPLC-NMR spectroscopic approaches have been used to quantify and identify the urinary-excreted metabolites of 4-trifluoromethoxyaniline (4-TFMeA) and its [13C]-labelled acetanilide following i.p. administration at 50 mg/kg to rats. The major metabolite excreted in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service