Skip to Content
Merck
2-Methyl-3-(trifluoromethyl)aniline 95%

2-Methyl-3-(trifluoromethyl)aniline

2-Methyl-3-(trifluoromethyl)aniline 95%
Synonym(s):
2-Amino-6-(trifluoromethyl)toluene
Linear Formula:
CH3C6H3(CF3)NH2
CAS Number:
Molecular Weight:
175.15
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing