Skip to Content
Merck
All Photos(2)

Key Documents

339121

Sigma-Aldrich

Zirconium(IV) ethoxide

97%

Synonym(s):

Tetraethoxyzirconium, Zirconium ethoxide (Zr(OEt)4 ), Zirconium tetraethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(OC2H5)4
CAS Number:
Molecular Weight:
271.47
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

reaction suitability

core: zirconium
reagent type: catalyst

bp

235 °C/5 mmHg (lit.)

SMILES string

CCO[Zr](OCC)(OCC)OCC

InChI

1S/4C2H5O.Zr/c4*1-2-3;/h4*2H2,1H3;/q4*-1;+4

InChI key

ASBGGHMVAMBCOR-UHFFFAOYSA-N

Application

Convenient precursor for the synthesis of olefin-functionalized zirconium-containing monomers, which have potential for the generation of hybrid zirconium-containing polymers.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William J. Evans et al.
Inorganic chemistry, 38(6), 1160-1164 (2001-10-24)
The attachment of pendent olefin groups to oxygen-ligated zirconium complexes using olefin-substituted phenols and alcohols and readily accessible zirconium reagents is described. Syntheses of three crystallographically characterizable complexes isolable in 55-90% yield are reported. Eugenol, HOC(6)H(3)(OMe-2)(CH(2)CH=CH(2)-4) (HOAr) reacts with [Zr(O(i)Pr)(4)(HO(i)Pr)](2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service