Skip to Content
Merck
All Photos(1)

Key Documents

337382

Sigma-Aldrich

Silver phosphate

98%

Synonym(s):

Silver orthophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag3PO4
CAS Number:
Molecular Weight:
418.58
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

mp

849 °C (lit.)

SMILES string

[Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O

InChI

1S/3Ag.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3

InChI key

FJOLTQXXWSRAIX-UHFFFAOYSA-K

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Wang et al.
Journal of the American Chemical Society, 134(9), 4096-4099 (2012-02-15)
A kinetic resolution of α-allenic alcohols is realized through chiral silver phosphate-catalyzed cycloisomerization with high stereoselectivity (selectivity factor up to 189) and tolerance of a variety of functional groups. A mechanistic model is proposed to interpret the origin of the
Stanislaw Halas et al.
Rapid communications in mass spectrometry : RCM, 25(5), 579-584 (2011-02-04)
Stable oxygen isotope compositions (δ(18)O values) of two commercial and one synthesized silver orthophosphate reagents have been determined on the VSMOW scale. The analyses were carried out in three different laboratories: lab (1) applying off-line oxygen extraction in the form
D F LaPorte et al.
Journal of mass spectrometry : JMS, 44(6), 879-890 (2009-01-09)
Oxygen isotope values of biogenic apatite have long demonstrated considerable promise for paleothermometry potential because of the abundance of material in the fossil record and greater resistance of apatite to diagenesis compared to carbonate. Unfortunately, this promise has not been
Wentao Duan et al.
Journal of the American Chemical Society, 135(4), 1280-1283 (2013-01-11)
We report a Ag(3)PO(4) microparticle system showing collective behaviors in aqueous medium. Transition between two emergent patterns, namely "exclusion" and "schooling", can be triggered by shift in chemical equilibrium upon the addition or removal of NH(3) or in response to
Y L Lou et al.
Journal of dentistry, 39(9), 612-618 (2011-07-13)
Silver diammine fluoride (SDF) is used as an anticaries agent; however, its mode of action is uncertain, whether chemical, physical, mechanical or antibacterial. As a preliminary study, the effect of SDF on hydroxyapatite (HAp) and gelatin (as a chemically-representative protein)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service