Skip to Content
Merck
All Photos(2)

Key Documents

226831

Sigma-Aldrich

Silver(I) oxide

≥99.99% trace metals basis

Synonym(s):

Argentous oxide, Disilver monoxide, Disilver oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag2O
CAS Number:
Molecular Weight:
231.74
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: silver

SMILES string

[Ag]O[Ag]

InChI

1S/2Ag.O

InChI key

KHJDQHIZCZTCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxidizing agent for the generation of lactones from diols. Mediates monoprotection of symmetrical diols with alkyl halides in good to excellent yield.
Stoichiometric quantities of Ag2O convert tertiary bromides to esters, ethers, and amines.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Serb. Chem. Soc., 53, 411-411 (1988)
Tetrahedron Letters, 48, 5761-5761 (2007)
Bouzide, A. Sauve, G.
Tetrahedron Letters, 38, 5945-5945 (1997)
Woonsup Shin et al.
Analytical chemistry, 83(12), 5023-5025 (2011-05-10)
When a current or a voltage is applied across the ceramic membrane of the nongassing Ag/Ag(2)O-SiO(2)-Ag/Ag(2)O pump, protons produced in the anodic reaction 2Ag(s) + H(2)O → Ag(2)O(s) + 2H(+) + 2e(-) are driven to the cathode, where they are
Zijie Yan et al.
Langmuir : the ACS journal of surfaces and colloids, 27(2), 851-855 (2010-12-18)
A new route to synthesis of Ag(2)O micro-/nanostructures, including a mixture of cubes, pyramids, triangular plates, pentagonal rods, and bars, has been developed by pulsed excimer laser ablation of bulk silver in water using polysorbate 80 as surfactant. The polysorbate

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service