Skip to Content
Merck
All Photos(2)

Key Documents

330485

Sigma-Aldrich

1-Bromoeicosane

≥97%

Synonym(s):

Eicosyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19Br
CAS Number:
Molecular Weight:
361.44
Beilstein:
1763647
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

12.5 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥97%

mp

36-39 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CCCCCCCCCCCCCCCCCCCCBr

InChI

1S/C20H41Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h2-20H2,1H3

InChI key

CZASMUMJSKOHFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Bromoeicosane was used in the preparation of CuBr/polymer nanocomposite.

Biochem/physiol Actions

Self-assembled monolayers of 1-bromoeicosane were evaluated at the vacuum-graphite and liquid-graphite interfaces using scanning tunneling microscopy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent effects on the self-assembly of 1-bromoeicosane on graphite. Part I. Scanning tunneling microscopy.
Florio GM, et al.
The Journal of Physical Chemistry C, 113(9), 3631-3640 (2009)
Laser co-photolytic approach to copper (I) bromide/polymer nanosol and nanocomposite.
Pola J, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 190(1), 29-33 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service